Assay Design



Gene Transcripts

Gene Symbol
twk-10
Entrez Gene ID
192080
Gene Name
protein-coding
Gene Aliases
CELE_K04A8.4
Chromosome Location
Chr.V: 6552340 - 6560256 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313205.2 NP_001300134.1 9 - 10 1038 65
NM_001377597.2 NP_001364744.1 1 - 2 94 65

View Full Product Details