Assay Design



Gene Transcripts

Gene Symbol
twk-10
Entrez Gene ID
192080
Gene Name
protein-coding
Gene Aliases
CELE_K04A8.4
Chromosome Location
Chr.V: 6552340 - 6560256 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313205.2 NP_001300134.1 10 - 11 1205 102
NM_001377597.2 NP_001364744.1 2 - 3 261 102
NM_001377598.1 NP_001364841.1 1 - 2 165 102

View Full Product Details