Assay Design
Gene Transcripts
- Gene Symbol
- scd-2
- Entrez Gene ID
-
179054
- Gene Name
- ALK tyrosine kinase receptor homolog scd-2
- Gene Aliases
-
CELE_T10H9.2
- Chromosome Location
-
Chr.V: 6633225 - 6639665 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072284.4 |
NP_504685.3 |
19 - 20 |
3447 |
95 |
View Full Product Details