Assay Design
Gene Transcripts
- Gene Symbol
- snb-1
- Entrez Gene ID
-
266648
- Gene Name
- Synaptobrevin-1
- Gene Aliases
-
CELE_T10H9.4
- Chromosome Location
-
Chr.V: 6655586 - 6656501 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383325.1 |
|
1 - 2 |
142 |
75 |
GenBank mRNA |
AF003281.1 |
- |
1 - 2 |
148 |
75 |
View Full Product Details