Assay Design



Gene Transcripts

Gene Symbol
unc-70
Entrez Gene ID
179077
Gene Name
protein-coding
Gene Aliases
CELE_K11C4.3
Chromosome Location
Chr.V: 6881338 - 6891839 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028881.4 NP_001024052.1 9 - 10 6537 69
NM_001028882.5 NP_001024053.2 11 - 12 6672 69
NM_001269144.3 NP_001256073.1 8 - 9 6753 69
NM_001307743.4 NP_001294672.1 9 - 10 6569 69
GenBank mRNA AF166169.1 - 10 - 11 6620 69
AF166170.1 - 8 - 9 6485 69
AF261891.1 - 9 - 10 6553 69

View Full Product Details