Assay Design



Gene Transcripts

Gene Symbol
npa-1
Entrez Gene ID
179095
Gene Name
protein-coding
Gene Aliases
CELE_VC5.3
Chromosome Location
Chr.V: 7075877 - 7081899 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373048.3 NP_001360011.1 9 - 10 3059 87
NM_072394.7 NP_504795.1 9 - 10 3098 87
GenBank mRNA AY519856.1 - 9 - 10 3040 87

View Full Product Details