Assay Design
Gene Transcripts
- Gene Symbol
- ugt-40
- Entrez Gene ID
-
184292
- Gene Name
- UDP-GlucuronosylTransferase
- Gene Aliases
-
CELE_F10D2.5
- Chromosome Location
-
Chr.V: 7139475 - 7141964 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072409.7 |
NP_504810.2 |
3 - 4 |
588 |
125 |
View Full Product Details