Assay Design
Gene Transcripts
- Gene Symbol
- ugt-41
- Entrez Gene ID
-
184295
- Gene Name
- UDP-GlucuronosylTransferase
- Gene Aliases
-
CELE_F10D2.11
- Chromosome Location
-
Chr.V: 7161068 - 7163595 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072417.5 |
NP_504818.2 |
5 - 6 |
751 |
113 |
View Full Product Details