Assay Design
Gene Transcripts
- Gene Symbol
- clec-7
- Entrez Gene ID
-
184314
- Gene Name
- C-type LECtin
- Gene Aliases
-
CELE_F10G2.3
- Chromosome Location
-
Chr.V: 7328392 - 7330002 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072464.5 |
NP_504865.1 |
3 - 4 |
495 |
101 |
View Full Product Details