Assay Design
Gene Transcripts
- Gene Symbol
- srsx-24
- Entrez Gene ID
-
182178
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_C03G6.16
- Chromosome Location
-
Chr.V: 7377897 - 7379484 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072486.1 |
NP_504887.1 |
1 - 2 |
181 |
97 |
View Full Product Details