Assay Design
Gene Transcripts
- Gene Symbol
- srxa-4
- Entrez Gene ID
-
184638
- Gene Name
- Serpentine Receptor, class XA
- Gene Aliases
-
CELE_F18E3.1
- Chromosome Location
-
Chr.V: 7420462 - 7421713 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072501.2 |
NP_504902.2 |
1 - 2 |
243 |
134 |
View Full Product Details