Assay Design
Gene Transcripts
- Gene Symbol
- try-8
- Entrez Gene ID
-
179134
- Gene Name
- Peptidase S1 domain-containing protein
- Gene Aliases
-
CELE_F25E5.10
- Chromosome Location
-
Chr.V: 7452912 - 7454336 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072515.3 |
NP_504916.1 |
2 - 3 |
602 |
69 |
View Full Product Details