Assay Design
Gene Transcripts
- Gene Symbol
- str-31
- Entrez Gene ID
-
179136
- Gene Name
- Seven TM Receptor
- Gene Aliases
-
CELE_C54F6.10
- Chromosome Location
-
Chr.V: 7506869 - 7508666 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072531.2 |
NP_504932.1 |
3 - 4 |
1334 |
90 |
View Full Product Details