Assay Design
Gene Transcripts
- Gene Symbol
- srb-18
- Entrez Gene ID
-
183806
- Gene Name
- Serpentine receptor class beta-18
- Gene Aliases
-
CELE_C54F6.7
- Chromosome Location
-
Chr.V: 7515239 - 7517353 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072534.4 |
NP_504935.3 |
3 - 4 |
547 |
144 |
View Full Product Details