Assay Design
Gene Transcripts
- Gene Symbol
- srw-3
- Entrez Gene ID
-
191221
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_ZK105.2
- Chromosome Location
-
Chr.V: 7705616 - 7706955 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072594.2 |
NP_504995.2 |
2 - 3 |
853 |
137 |
View Full Product Details