Assay Design
Gene Transcripts
- Gene Symbol
- cyp-32A1
- Entrez Gene ID
-
179156
- Gene Name
- CYtochrome P450 family
- Gene Aliases
-
CELE_C26F1.2
- Chromosome Location
-
Chr.V: 7781050 - 7784595 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072608.11 |
NP_505009.4 |
5 - 6 |
711 |
122 |
View Full Product Details