Assay Design



Gene Transcripts

Gene Symbol
srsx-7
Entrez Gene ID
190819
Gene Name
Serpentine Receptor, class SX
Gene Aliases
CELE_Y97E10B.4
Chromosome Location
Chr.V: 7925783 - 7926920 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001368455.1 NP_001355388.1 1 - 2 138 128

View Full Product Details