Assay Design
Gene Transcripts
- Gene Symbol
- srsx-5
- Entrez Gene ID
-
190817
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_Y97E10B.2
- Chromosome Location
-
Chr.V: 7928960 - 7930416 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072637.3 |
NP_505038.2 |
1 - 2 |
129 |
134 |
View Full Product Details