Assay Design
Gene Transcripts
- Gene Symbol
- sdz-23
- Entrez Gene ID
-
186544
- Gene Name
- EGF-like domain-containing protein
- Gene Aliases
-
CELE_F58G4.4
- Chromosome Location
-
Chr.V: 8099606 - 8100513 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072690.2 |
NP_505091.1 |
1 - 2 |
498 |
119 |
View Full Product Details