Assay Design



Gene Transcripts

Gene Symbol
spr-1
Entrez Gene ID
179192
Gene Name
REST corepressor spr-1
Gene Aliases
CELE_D1014.8
Chromosome Location
Chr.V: 8126663 - 8129352 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072697.5 NP_505098.1 7 - 8 1115 128
GenBank mRNA AF548624.1 - 7 - 8 1115 128
AY160228.1 - 7 - 8 1112 128

View Full Product Details