Assay Design
Gene Transcripts
- Gene Symbol
- snap-1
- Entrez Gene ID
-
179193
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_D1014.3
- Chromosome Location
-
Chr.V: 8130613 - 8133334 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072698.6 |
NP_505099.2 |
2 - 3 |
134 |
74 |
View Full Product Details