Assay Design



Gene Transcripts

Gene Symbol
snap-1
Entrez Gene ID
179193
Gene Name
protein-coding
Gene Aliases
CELE_D1014.3
Chromosome Location
Chr.V: 8130613 - 8133334 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373059.3 NP_001360536.1 - 970 88
NM_072698.6 NP_505099.2 3 - 4 297 88

View Full Product Details