Assay Design
Gene Transcripts
- Gene Symbol
- air-1
- Entrez Gene ID
-
179202
- Gene Name
- Protein kinase domain-containing protein
- Gene Aliases
-
CELE_K07C11.2
- Chromosome Location
-
Chr.V: 8221488 - 8223585 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072718.10 |
NP_505119.1 |
4 - 5 |
459 |
81 |
GenBank mRNA |
AF071206.1 |
- |
4 - 5 |
476 |
81 |
View Full Product Details