Assay Design



Gene Transcripts

Gene Symbol
acr-16
Entrez Gene ID
179235
Gene Name
Acetylcholine receptor subunit alpha-type acr-16
Gene Aliases
CELE_F25G6.3
Chromosome Location
Chr.V: 8561216 - 8564345 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072806.5 NP_505207.1 5 - 6 746 103
GenBank mRNA AY523511.1 - 5 - 6 746 103
X83887.1 - - 733 103

View Full Product Details