Assay Design
Gene Transcripts
- Gene Symbol
- oac-29
- Entrez Gene ID
-
185622
- Gene Name
- Acyl_transf_3 domain-containing protein
- Gene Aliases
-
CELE_F41E6.14
- Chromosome Location
-
Chr.V: 8631871 - 8635675 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072827.3 |
NP_505228.2 |
4 - 5 |
925 |
70 |
View Full Product Details