Assay Design
Gene Transcripts
- Gene Symbol
- glr-5
- Entrez Gene ID
-
179251
- Gene Name
- GLutamate Receptor family (AMPA)
- Gene Aliases
-
CELE_ZC196.7
- Chromosome Location
-
Chr.V: 8716495 - 8721036 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072843.5 |
NP_505244.1 |
5 - 6 |
1258 |
141 |
View Full Product Details