Assay Design



Gene Transcripts

Gene Symbol
gck-1
Entrez Gene ID
179274
Gene Name
protein-coding
Gene Aliases
CELE_T19A5.2
Chromosome Location
Chr.V: 8947799 - 8956296 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072909.5 NP_505310.2 1 - 2 537 75
NM_072910.8 NP_505311.3 1 - 2 537 75

View Full Product Details