Assay Design



Gene Transcripts

Gene Symbol
oac-50
Entrez Gene ID
179284
Gene Name
NRF domain-containing protein
Gene Aliases
CELE_T27E4.6
Chromosome Location
Chr.V: 9069857 - 9073540 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322580.2 NP_001309487.1 2 - 3 287 149

View Full Product Details