Assay Design
Gene Transcripts
- Gene Symbol
- glo-4
- Entrez Gene ID
-
179306
- Gene Name
- X-linked retinitis pigmentosa GTPase regulator homolog
- Gene Aliases
-
CELE_F07C3.4
- Chromosome Location
-
Chr.V: 9234610 - 9243221 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072991.5 |
NP_505392.3 |
8 - 9 |
1500 |
102 |
View Full Product Details