Assay Design



Gene Transcripts

Gene Symbol
nhr-36
Entrez Gene ID
24104398
Gene Name
Nuclear Hormone Receptor family
Gene Aliases
CELE_F07C3.10
Chromosome Location
Chr.V: 9258136 - 9261592 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307788.5 NP_001294717.2 7 - 8 1387 82

View Full Product Details