Assay Design
Gene Transcripts
- Gene Symbol
- ucp-4
- Entrez Gene ID
-
179315
- Gene Name
- UnCoupling Protein (mitochondrial substrate carrier)
- Gene Aliases
-
CELE_K07B1.3
- Chromosome Location
-
Chr.V: 9334298 - 9336671 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073013.7 |
NP_505414.1 |
2 - 3 |
138 |
69 |
View Full Product Details