Assay Design
Gene Transcripts
- Gene Symbol
- hcp-2
- Entrez Gene ID
-
179353
- Gene Name
- HoloCentric chromosome binding Protein
- Gene Aliases
-
CELE_T06E4.1
- Chromosome Location
-
Chr.V: 9645746 - 9650211 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073088.4 |
NP_505489.1 |
7 - 8 |
3807 |
78 |
View Full Product Details