Assay Design
Gene Transcripts
- Gene Symbol
- cyp-29A4
- Entrez Gene ID
-
181926
- Gene Name
- CYtochrome P450 family
- Gene Aliases
-
CELE_B0331.1
- Chromosome Location
-
Chr.V: 9654755 - 9658120 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073089.4 |
NP_505490.2 |
5 - 6 |
660 |
87 |
View Full Product Details