Assay Design
Gene Transcripts
- Gene Symbol
- ztf-7
- Entrez Gene ID
-
179375
- Gene Name
- C2H2-type domain-containing protein
- Gene Aliases
-
CELE_F46B6.7
- Chromosome Location
-
Chr.V: 9786949 - 9789701 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073125.6 |
NP_505526.1 |
2 - 3 |
383 |
64 |
View Full Product Details