Assay Design



Gene Transcripts

Gene Symbol
sru-39
Entrez Gene ID
185842
Gene Name
protein-coding
Gene Aliases
CELE_F46B6.11
Chromosome Location
Chr.V: 9803476 - 9804634 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276917.4 NP_001263846.1 3 - 4 703 132
NM_073129.3 NP_505530.1 3 - 4 706 132

View Full Product Details