Assay Design
Gene Transcripts
- Gene Symbol
- srsx-21
- Entrez Gene ID
-
187652
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_R07B5.5
- Chromosome Location
-
Chr.V: 9837979 - 9839508 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073138.5 |
NP_505539.2 |
1 - 2 |
123 |
69 |
View Full Product Details