Assay Design



Gene Transcripts

Gene Symbol
ric-4
Entrez Gene ID
179427
Gene Name
protein-coding
Gene Aliases
CELE_Y22F5A.3
Chromosome Location
Chr.V: 10261127 - 10270314 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129561.4 NP_001123033.1 4 - 5 720 70
NM_073240.8 NP_505641.2 4 - 5 589 70

View Full Product Details