Assay Design



Gene Transcripts

Gene Symbol
gcy-6
Entrez Gene ID
191644
Gene Name
protein-coding
Gene Aliases
CELE_B0024.6
Chromosome Location
Chr.V: 10308291 - 10314053 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307767.1 NP_001294696.1 11 - 12 1541 112
NM_073249.3 NP_505650.3 11 - 12 1529 112

View Full Product Details