Assay Design



Gene Transcripts

Gene Symbol
ceeh-2
Entrez Gene ID
179444
Gene Name
Epoxide hydrolase 2
Gene Aliases
CELE_K07C5.5
Chromosome Location
Chr.V: 10356285 - 10357903 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269282.2 NP_001256211.1 3 - 4 1020 106
GenBank mRNA EU151492.1 - 3 - 4 782 106

View Full Product Details