Assay Design



Gene Transcripts

Gene Symbol
cash-1
Entrez Gene ID
179445
Gene Name
protein-coding
Gene Aliases
CELE_K07C5.8
Chromosome Location
Chr.V: 10362261 - 10365964 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373096.3 NP_001360512.1 7 - 8 1433 111
NM_073263.5 NP_505664.2 7 - 8 1424 111

View Full Product Details