Assay Design
Gene Transcripts
- Gene Symbol
- ugt-2
- Entrez Gene ID
-
179450
- Gene Name
- UDP-GlucuronosylTransferase
- Gene Aliases
-
CELE_AC3.8
- Chromosome Location
-
Chr.V: 10398717 - 10401307 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073271.3 |
NP_505672.2 |
10 - 11 |
1515 |
118 |
View Full Product Details