Assay Design
Gene Transcripts
- Gene Symbol
- snf-10
- Entrez Gene ID
-
191769
- Gene Name
- Transporter
- Gene Aliases
-
CELE_Y32F6A.2
- Chromosome Location
-
Chr.V: 10438330 - 10440823 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073281.4 |
NP_505682.2 |
8 - 9 |
1952 |
113 |
View Full Product Details