Assay Design



Gene Transcripts

Gene Symbol
scpl-4
Entrez Gene ID
179481
Gene Name
protein-coding
Gene Aliases
CELE_T21C9.12
Chromosome Location
Chr.V: 10599104 - 10602147 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373099.3 NP_001360575.1 1 - 2 79 68
NM_073321.6 NP_505722.1 1 - 2 78 68

View Full Product Details