Assay Design
Gene Transcripts
- Gene Symbol
- ugt-34
- Entrez Gene ID
-
185120
- Gene Name
- UDP-GlucuronosylTransferase
- Gene Aliases
-
CELE_F29F11.2
- Chromosome Location
-
Chr.V: 10654415 - 10656978 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073327.6 |
NP_505728.2 |
3 - 4 |
748 |
118 |
View Full Product Details