Assay Design



Gene Transcripts

Gene Symbol
gsp-1
Entrez Gene ID
179486
Gene Name
Serine/threonine-protein phosphatase PP1-alpha
Gene Aliases
CELE_F29F11.6
Chromosome Location
Chr.V: 10676261 - 10686506 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269321.3 NP_001256250.1 6 - 7 1364 101

View Full Product Details