Assay Design



Gene Transcripts

Gene Symbol
nas-31
Entrez Gene ID
186493
Gene Name
protein-coding
Gene Aliases
CELE_F58B4.1
Chromosome Location
Chr.V: 10918857 - 10921847 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028822.4 NP_001023993.1 2 - 3 365 86
NM_001028823.6 NP_001023994.1 2 - 3 343 86

View Full Product Details