Assay Design
Gene Transcripts
- Gene Symbol
- srg-41
- Entrez Gene ID
-
188592
- Gene Name
- Serpentine receptor class gamma
- Gene Aliases
-
CELE_T19C4.3
- Chromosome Location
-
Chr.V: 11160452 - 11161891 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073436.1 |
NP_505837.1 |
1 - 2 |
245 |
83 |
View Full Product Details