Assay Design
Gene Transcripts
- Gene Symbol
- srg-40
- Entrez Gene ID
-
188593
- Gene Name
- Serpentine receptor class gamma
- Gene Aliases
-
CELE_T19C4.4
- Chromosome Location
-
Chr.V: 11163206 - 11164575 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073437.4 |
NP_505838.2 |
3 - 4 |
510 |
97 |
View Full Product Details