Assay Design



Gene Transcripts

Gene Symbol
gpa-1
Entrez Gene ID
179546
Gene Name
protein-coding
Gene Aliases
CELE_T19C4.6
Chromosome Location
Chr.V: 11174433 - 11177677 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129546.4 NP_001123018.1 5 - 6 729 69
NM_073439.3 NP_505840.1 7 - 8 1003 69
GenBank mRNA AY008124.1 - 7 - 8 885 69

View Full Product Details