Assay Design
Gene Transcripts
- Gene Symbol
- nas-17
- Entrez Gene ID
-
186923
- Gene Name
- Zinc metalloproteinase nas-17
- Gene Aliases
-
CELE_K03B8.2
- Chromosome Location
-
Chr.V: 11398916 - 11400563 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073491.2 |
NP_505892.1 |
2 - 3 |
495 |
65 |
View Full Product Details